Assay Design



Gene Transcripts

Gene Symbol
Arl5a
Entrez Gene ID
117050
Gene Name
ADP-ribosylation factor-like 5A
Gene Aliases
Arl5
Chromosome Location
Chr.3: 37922544 - 37947434 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053979.2 NP_446431.1 3 - 4 453 83
XM_008761696.1 XP_008759918.1 3 - 4 525 83
GenBank mRNA BC097294.1 - 3 - 4 453 83
X78604.1 - 3 - 4 302 83

View Full Product Details