Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012881.2 | NP_037013.2 | 6 - 7 | 598 | 124 |
XM_008769996.1 | XP_008768218.1 | 6 - 7 | 671 | 124 | |
GenBank mRNA | AB001382.1 | - | 5 - 6 | 492 | 124 |
BC078874.1 | - | 6 - 7 | 578 | 124 | |
FQ213473.1 | - | 6 - 7 | 587 | 124 | |
FQ222192.1 | - | 6 - 7 | 614 | 124 | |
M14656.1 | - | 6 - 7 | 571 | 124 | |
M99252.1 | - | 6 - 7 | 607 | 124 |