Assay Design



Gene Transcripts

Gene Symbol
Glt8d1
Entrez Gene ID
306253
Gene Name
glycosyltransferase 8 domain containing 1
Gene Aliases
Da2-24
Chromosome Location
Chr.16: 7082450 - 7097405 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007683.1 NP_001007684.1 7 - 8 823 128
GenBank mRNA AY325256.1 - 7 - 8 770 128
BC079066.1 - 7 - 8 823 128

View Full Product Details