Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286936.1 | NP_001273865.1 | 4 - 5 | 521 | 118 |
XM_006230901.2 | XP_006230963.1 | 4 - 5 | 497 | 118 | |
XM_008760175.1 | XP_008758397.1 | 4 - 5 | 485 | 118 | |
GenBank mRNA | DY546174.1 | - | 2 - 3 | 241 | 118 |
FQ227385.1 | - | 4 - 5 | 557 | 118 |