Assay Design
Gene Transcripts
- Gene Symbol
- Fgf13
- Entrez Gene ID
-
84488
- Gene Name
- fibroblast growth factor 13
- Gene Aliases
-
FGF-13
- Chromosome Location
-
Chr.X: 142053648 - 142571077 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053428.1 |
NP_445880.1 |
1 - 2 |
146 |
135 |
GenBank mRNA |
AF271786.1 |
- |
1 - 2 |
146 |
135 |
View Full Product Details