Assay Design



Gene Transcripts

Gene Symbol
Pdia4
Entrez Gene ID
116598
Gene Name
protein disulfide isomerase family A, member 4
Gene Aliases
ERp-72, Erp70, Erp72
Chromosome Location
Chr.4: 77470636 - 77489535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053849.1 NP_446301.1 9 - 10 1806 95
GenBank mRNA BC061535.1 - 9 - 10 1620 95
M86870.1 - 9 - 10 1806 95

View Full Product Details