Assay Design



Gene Transcripts

Gene Symbol
Vopp1
Entrez Gene ID
362374
Gene Name
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases
Ecop, RGD1306494
Chromosome Location
Chr.4: 88441067 - 88515141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108630.1 NP_001102100.1 1 - 2 207 127
GenBank mRNA BC168709.1 - 1 - 2 200 127

View Full Product Details