Assay Design



Gene Transcripts

Gene Symbol
Rsrc1
Entrez Gene ID
361956
Gene Name
arginine/serine-rich coiled-coil 1
Gene Aliases
RGD1304968, SRrp53
Chromosome Location
Chr.2: 164126898 - 164533652 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014172.1 NP_001014194.1 9 - 10 1061 79
GenBank mRNA BC087665.1 - 9 - 10 1061 79

View Full Product Details