Assay Design



Gene Transcripts

Gene Symbol
Stradb
Entrez Gene ID
501146
Gene Name
STE20-related kinase adaptor beta
Gene Aliases
Als2cr2, RGD1559449
Chromosome Location
Chr.9: 65763437 - 65782513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109307.2 NP_001102777.2 7 - 8 906 68

View Full Product Details