Assay Design



Gene Transcripts

Gene Symbol
Tcf3
Entrez Gene ID
171046
Gene Name
transcription factor 3
Gene Aliases
Pan2, Tcfe2a
Chromosome Location
Chr.7: 12146642 - 12168400 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035237.1 NP_001030314.1 6 - 7 574 96
NM_133524.2 NP_598208.2 6 - 7 574 96
GenBank mRNA AJ973227.1 - 6 - 7 574 96
BC078750.1 - 6 - 7 502 96
X54549.1 - 6 - 7 574 96

View Full Product Details