Assay Design



Gene Transcripts

Gene Symbol
Nfe2l1
Entrez Gene ID
360610
Gene Name
nuclear factor, erythroid 2-like 1
Gene Aliases
-
Chromosome Location
Chr.10: 84686762 - 84699301 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247236.2 XP_006247298.1 2 - 3 1114 67
XM_006247237.2 XP_006247299.1 2 - 3 1106 67
GenBank mRNA BC169095.1 - - 508 67

View Full Product Details