Assay Design



Gene Transcripts

Gene Symbol
Aoc3
Entrez Gene ID
29473
Gene Name
amine oxidase, copper containing 3
Gene Aliases
SSAO
Chromosome Location
Chr.10: 89251370 - 89259313 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031582.2 NP_113770.2 1 - 2 1676 60
GenBank mRNA AB195675.1 - 1 - 2 1681 60
BC100613.1 - 1 - 2 1676 60

View Full Product Details