Assay Design



Gene Transcripts

Gene Symbol
Tsen54
Entrez Gene ID
690308
Gene Name
TSEN54 tRNA splicing endonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.10: 104358419 - 104366926 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109576.1 NP_001103046.1 3 - 4 343 140
XM_006247737.2 XP_006247799.1 3 - 4 1266 140
GenBank mRNA BC166577.1 - 3 - 4 287 140

View Full Product Details