Assay Design



Gene Transcripts

Gene Symbol
Tbrg4
Entrez Gene ID
360977
Gene Name
transforming growth factor beta regulator 4
Gene Aliases
-
Chromosome Location
Chr.14: 86876800 - 86885604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251342.2 XP_006251404.1 1 - 2 106 109
GenBank mRNA FQ217643.1 - 1 - 2 69 109
FQ217828.1 - 1 - 2 69 109

View Full Product Details