Assay Design



Gene Transcripts

Gene Symbol
Eif3b
Entrez Gene ID
288516
Gene Name
eukaryotic translation initiation factor 3, subunit B
Gene Aliases
Eif3s9
Chromosome Location
Chr.12: 16285730 - 16310190 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031640.1 NP_001026810.1 15 - 16 2130 154
GenBank mRNA BC098728.1 - 15 - 16 2130 154

View Full Product Details