Assay Design



Gene Transcripts

Gene Symbol
Grifin
Entrez Gene ID
117130
Gene Name
galectin-related inter-fiber protein
Gene Aliases
-
Chromosome Location
Chr.12: 16170108 - 16172144 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057187.2 NP_476535.1 2 - 3 100 80
XM_008768938.1 XP_008767160.1 2 - 3 154 80
GenBank mRNA AF082160.1 - 2 - 3 100 80

View Full Product Details