Assay Design



Gene Transcripts

Gene Symbol
Uspl1
Entrez Gene ID
288447
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
RGD1310835
Chromosome Location
Chr.12: 6930018 - 6957026 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105906.2 NP_001099376.1 8 - 9 1558 87
NM_001198555.1 NP_001185484.1 8 - 9 1558 87
XM_006248810.2 XP_006248872.1 7 - 8 1501 87
GenBank mRNA CO396482.1 - 4 - 5 471 87
EV774129.1 - 3 - 4 366 87

View Full Product Details