Assay Design



Gene Transcripts

Gene Symbol
Uspl1
Entrez Gene ID
288447
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
RGD1310835
Chromosome Location
Chr.12: 6930018 - 6957026 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105906.2 NP_001099376.1 1 - 2 129 132
NM_001198555.1 NP_001185484.1 1 - 2 129 132
GenBank mRNA CB787426.1 - 1 - 2 141 132

View Full Product Details