Assay Design



Gene Transcripts

Gene Symbol
Eif2ak1
Entrez Gene ID
27137
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 1
Gene Aliases
Hri
Chromosome Location
Chr.12: 12749026 - 12782078 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013223.1 NP_037355.1 2 - 3 293 67
GenBank mRNA BC081838.1 - 2 - 3 326 67
L27707.1 - 2 - 3 293 67

View Full Product Details