Assay Design



Gene Transcripts

Gene Symbol
Mrpl23
Entrez Gene ID
64360
Gene Name
mitochondrial ribosomal protein L23
Gene Aliases
Rpl23-rs, Rpl23l, rL23MRP
Chromosome Location
Chr.1: 215701544 - 215709313 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022529.1 NP_071974.1 4 - 5 309 81
GenBank mRNA BC059936.1 - 4 - 5 315 81
U62635.1 - 4 - 5 309 81

View Full Product Details