Assay Design
Gene Transcripts
- Gene Symbol
- Zp1
- Entrez Gene ID
-
85271
- Gene Name
- zona pellucida glycoprotein 1 (sperm receptor)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 226966366 - 226972746 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053509.1 |
NP_445961.1 |
5 - 6 |
1019 |
62 |
GenBank mRNA |
AB000928.1 |
- |
5 - 6 |
1019 |
62 |
View Full Product Details