Assay Design



Gene Transcripts

Gene Symbol
Pank1
Entrez Gene ID
294088
Gene Name
pantothenate kinase 1
Gene Aliases
-
Chromosome Location
Chr.1: 253115400 - 253189644 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106373.3 NP_001099843.1 4 - 5 979 86
XM_006231276.2 XP_006231338.1 4 - 5 4282 86
XM_006231278.2 XP_006231340.1 5 - 6 1630 86

View Full Product Details