Assay Design



Gene Transcripts

Gene Symbol
Cbwd1
Entrez Gene ID
171057
Gene Name
COBW domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 242819383 - 242861805 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133535.2 NP_598219.2 13 - 14 955 139
XM_006231191.2 XP_006231253.1 12 - 13 974 139
XM_006231192.2 XP_006231254.1 11 - 12 941 139
XM_006231193.2 XP_006231255.1 10 - 11 872 139
XM_008760303.1 XP_008758525.1 12 - 13 986 139
GenBank mRNA AF353305.1 - 13 - 14 1011 139
BC086376.1 - 13 - 14 955 139

View Full Product Details