Assay Design



Gene Transcripts

Gene Symbol
Cd81
Entrez Gene ID
25621
Gene Name
Cd81 molecule
Gene Aliases
Tapa1
Chromosome Location
Chr.1: 216237697 - 216253311 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013087.1 NP_037219.1 5 - 6 687 63
GenBank mRNA BC060583.1 - 5 - 6 63
FQ229194.1 - 3 - 4 267 63
FQ231991.1 - 5 - 6 673 63
FQ232886.1 - 5 - 6 675 63
U19894.1 - 5 - 6 687 63

View Full Product Details