Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013087.1 | NP_037219.1 | 7 - 8 | 867 | 90 |
GenBank mRNA | BC060583.1 | - | 7 - 8 | 698 | 90 |
FQ229194.1 | - | 5 - 6 | 447 | 90 | |
FQ231991.1 | - | 7 - 8 | 853 | 90 | |
FQ232886.1 | - | 7 - 8 | 856 | 91 | |
U19894.1 | - | 7 - 8 | 867 | 90 |