Assay Design



Gene Transcripts

Gene Symbol
Npas4
Entrez Gene ID
266734
Gene Name
neuronal PAS domain protein 4
Gene Aliases
Nxf
Chromosome Location
Chr.1: 220260310 - 220278024 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153626.1 NP_705890.1 3 - 4 467 78
XM_006230697.2 XP_006230759.1 5 - 6 2624 78
GenBank mRNA AB050103.1 - 3 - 4 467 78

View Full Product Details