Assay Design
Gene Transcripts
- Gene Symbol
- Gipr
- Entrez Gene ID
-
25024
- Gene Name
- gastric inhibitory polypeptide receptor
- Gene Aliases
-
Gippr, RATGIPPR
- Chromosome Location
-
Chr.1: 80063049 - 80073223 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012714.1 |
NP_036846.1 |
11 - 12 |
1173 |
96 |
GenBank mRNA |
L19660.1 |
- |
11 - 12 |
1173 |
96 |
View Full Product Details