Assay Design



Gene Transcripts

Gene Symbol
Tyrp1
Entrez Gene ID
298182
Gene Name
tyrosinase-related protein 1
Gene Aliases
B
Chromosome Location
Chr.5: 98387291 - 98406083 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106664.1 NP_001100134.1 2 - 3 625 87
XM_006238332.2 XP_006238394.1 2 - 3 656 87

View Full Product Details