Assay Design
Gene Transcripts
- Gene Symbol
- Shox2
- Entrez Gene ID
-
25546
- Gene Name
- short stature homeobox 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 164118175 - 164126783 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ002258.1 |
- |
4 - 5 |
455 |
124 |
View Full Product Details