Assay Design



Gene Transcripts

Gene Symbol
Fcrl2
Entrez Gene ID
310694
Gene Name
Fc receptor-like 2
Gene Aliases
Fcrls, Msr2
Chromosome Location
Chr.2: 186594442 - 186605115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107702.1 NP_001101172.1 4 - 5 1036 93

View Full Product Details