Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106450.1 | NP_001099920.1 | 3 - 4 | 417 | 93 |
XM_006232620.2 | XP_006232682.1 | 4 - 5 | 508 | 93 | |
XM_006232621.2 | XP_006232683.1 | 4 - 5 | 1125 | 93 | |
XM_006232622.1 | XP_006232684.1 | 3 - 4 | 675 | 93 | |
XM_008761141.1 | XP_008759363.1 | 4 - 5 | 827 | 93 |