Assay Design



Gene Transcripts

Gene Symbol
Slc27a3
Entrez Gene ID
295219
Gene Name
solute carrier family 27 (fatty acid transporter), member 3
Gene Aliases
-
Chromosome Location
Chr.2: 189760747 - 189765415 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106439.1 NP_001099909.1 3 - 4 1142 72

View Full Product Details