Assay Design



Gene Transcripts

Gene Symbol
Creb3l2
Entrez Gene ID
362339
Gene Name
cAMP responsive element binding protein 3-like 2
Gene Aliases
Ra69, SCIRR69
Chromosome Location
Chr.4: 64869285 - 64981519 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012188.1 NP_001012188.1 1 - 2 438 80
GenBank mRNA AY546001.1 - 1 - 2 438 80

View Full Product Details