Assay Design



Gene Transcripts

Gene Symbol
Col28a1
Entrez Gene ID
312115
Gene Name
collagen, type XXVIII, alpha 1
Gene Aliases
RGD1564680
Chromosome Location
Chr.4: 34048074 - 34198658 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749699.3 XP_003749747.1 34 - 35 3619 117

View Full Product Details