Assay Design



Gene Transcripts

Gene Symbol
Zswim7
Entrez Gene ID
287388
Gene Name
zinc finger, SWIM-type containing 7
Gene Aliases
RGD1310324
Chromosome Location
Chr.10: 48587117 - 48599208 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163492.1 NP_001156964.1 1 - 2 93 153
GenBank mRNA BF522197.1 - 1 - 2 100 153

View Full Product Details