Assay Design



Gene Transcripts

Gene Symbol
Dnajc19
Entrez Gene ID
502525
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 19
Gene Aliases
RGD1560220
Chromosome Location
Chr.2: 120504115 - 120531782 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134640.1 NP_001128112.1 3 - 4 268 101
XM_006232259.2 XP_006232321.1 3 - 4 283 101
GenBank mRNA FQ212146.1 - 3 - 4 227 101

View Full Product Details