Assay Design



Gene Transcripts

Gene Symbol
Sh2d1a
Entrez Gene ID
501502
Gene Name
SH2 domain containing 1A
Gene Aliases
RGD1562408
Chromosome Location
Chr.X: 128897042 - 128925408 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109313.2 NP_001102783.2 3 - 4 560 64
XM_008773528.1 XP_008771750.1 3 - 4 690 64
GenBank mRNA BC167036.1 - 3 - 4 560 64

View Full Product Details