Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108813.2 | NP_001102283.2 | 2 - 3 | 274 | 118 |
GenBank mRNA | CB567280.1 | - | 2 - 3 | 291 | 118 |
FM054992.1 | - | 2 - 3 | 259 | 118 | |
FQ214979.1 | - | 2 - 3 | 235 | 118 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008773489.1 | XP_008771711.1 | 2 - 3 | 365 | 118 |