Assay Design



Gene Transcripts

Gene Symbol
Prodh2
Entrez Gene ID
361538
Gene Name
proline dehydrogenase (oxidase) 2
Gene Aliases
-
Chromosome Location
Chr.1: 88955125 - 88968501 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038588.1 NP_001033677.1 7 - 8 1017 71
GenBank mRNA AF222852.1 - 7 - 8 1007 71
BC158806.1 - 7 - 8 1017 71

View Full Product Details