Assay Design



Gene Transcripts

Gene Symbol
Prodh2
Entrez Gene ID
361538
Gene Name
proline dehydrogenase (oxidase) 2
Gene Aliases
-
Chromosome Location
Chr.1: 88955125 - 88968501 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038588.1 NP_001033677.1 5 - 6 689 92
GenBank mRNA AF222852.1 - 5 - 6 679 92
BC158806.1 - 5 - 6 689 92

View Full Product Details