Assay Design
Gene Transcripts
- Gene Symbol
- Lhb
- Entrez Gene ID
-
25329
- Gene Name
- luteinizing hormone beta polypeptide
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 101410021 - 101413725 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U25802.1 |
- |
5 - 6 |
2062 |
99 |
View Full Product Details