Assay Design



Gene Transcripts

Gene Symbol
Ptov1
Entrez Gene ID
292888
Gene Name
prostate tumor overexpressed 1
Gene Aliases
-
Chromosome Location
Chr.1: 100859346 - 100865975 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008304.1 NP_001008305.1 5 - 6 671 65
GenBank mRNA BC085836.1 - 5 - 6 671 65

View Full Product Details