Assay Design



Gene Transcripts

Gene Symbol
Cd22
Entrez Gene ID
308501
Gene Name
CD22 molecule
Gene Aliases
-
Chromosome Location
Chr.1: 89313475 - 89329900 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107503.1 NP_001100973.1 1 - 2 334 65

View Full Product Details