Assay Design
Gene Transcripts
- Gene Symbol
- Mup5
- Entrez Gene ID
-
298107
- Gene Name
- major urinary protein 5
- Gene Aliases
-
PGCL6
- Chromosome Location
-
Chr.5: 77549993 - 77749613 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB039828.1 |
- |
- |
453 |
122 |
View Full Product Details