Assay Design
Gene Transcripts
- Gene Symbol
- Btg3
- Entrez Gene ID
-
54230
- Gene Name
- BTG family, member 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 16873642 - 16889100 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC127498.1 |
- |
1 - 2 |
128 |
126 |
View Full Product Details