Assay Design



Gene Transcripts

Gene Symbol
Btg3
Entrez Gene ID
54230
Gene Name
BTG family, member 3
Gene Aliases
-
Chromosome Location
Chr.11: 16873642 - 16889100 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019290.1 NP_062163.1 4 - 5 568 135
GenBank mRNA AF087037.1 - 4 - 5 568 135
BC127498.1 - 4 - 5 661 135

View Full Product Details