Assay Design



Gene Transcripts

Gene Symbol
Adamts5
Entrez Gene ID
304135
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif, 5
Gene Aliases
-
Chromosome Location
Chr.11: 25411174 - 25456639 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198761.1 NP_942056.1 2 - 3 1235 75
GenBank mRNA AY382879.1 - 2 - 3 1235 75
FQ228283.1 - - 1628 75

View Full Product Details