Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173558.1 | NP_001167029.1 | 9 - 10 | 1150 | 112 |
XM_006241605.2 | XP_006241667.1 | 9 - 10 | 1130 | 112 | |
GenBank mRNA | BC158730.1 | - | 7 - 8 | 744 | 112 |
FQ228874.1 | - | 9 - 10 | 1159 | 112 | |
FQ229670.1 | - | 7 - 8 | 809 | 112 |