Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173558.1 | NP_001167029.1 | 10 - 11 | 1318 | 129 |
XM_006241605.2 | XP_006241667.1 | 10 - 11 | 1298 | 129 | |
GenBank mRNA | BC158730.1 | - | 8 - 9 | 912 | 129 |
FQ228874.1 | - | 10 - 11 | 1327 | 129 | |
FQ229670.1 | - | 8 - 9 | 977 | 129 |