Assay Design



Gene Transcripts

Gene Symbol
Trim2
Entrez Gene ID
361970
Gene Name
tripartite motif-containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 183145096 - 183223819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108552.1 NP_001102022.1 7 - 8 1814 87

View Full Product Details